Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1-Tris(hydroxymethyl)ethane, 97%

Catalog No. p-209075
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 77-85-0 | C5H12O3 | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-85-0
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00004687
InChI Key QXJQHYBHAIHNGG-UHFFFAOYSA-N
Synonym trimethylolethane, 1,1,1-tris hydroxymethyl ethane, trimet, methriol, metriol, pentaglycerine, pentaglycerol, methyltrimethanolmethane, 2-hydroxymethyl-2-methylpropane-1,3-diol, tris hydroxymethyl ethane
PubChem CID 6502
IUPAC Name 2-(hydroxymethyl)-2-methylpropane-1,3-diol
SMILES CC(CO)(CO)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 200.0°C to 203.0°C
Color White
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C(CH2OH)3
Quantity 25 g
Beilstein 01, 520
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols, insoluble in ether, benzene
Formula Weight 120.15
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1, 1, 1-Tris(hydroxymethyl)ethane, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective g
EINECSNumber 201-063-9
RTECSNumber TY7137500
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.