Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2,3,3-Hexafluoro-1-(trifluoromethoxy)-3-[(1,2,2-trifluorovinyl)oxy]propane 97.0+%, TCI America™
SDP

Catalog No. H161025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H161025G 25 g
H16105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H161025G Supplier TCI America Supplier No. H161025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40573-09-9
Molecular Formula C8F16O5
Molecular Weight (g/mol) 480.06
MDL Number MFCD00506884
InChI Key HCGYVDVMTQESCQ-UHFFFAOYSA-N
PubChem CID 57349774
IUPAC Name hexadecafluoro-2,4,6,8,11-pentaoxatridec-12-ene
SMILES FC(F)=C(F)OC(F)(F)C(F)(F)OC(F)(F)OC(F)(F)OC(F)(F)OC(F)(F)F

Specifications

Color Colorless
Boiling Point 64°C
Quantity 25 g
Formula Weight 332.04
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2,2,3,3-Hexafluoro-1-(trifluoromethoxy)-3-[(1,2,2-trifluorovinyl)oxy]propane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.