Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrabromoethane, 98%

Catalog No. p-3830540
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle

CAS: 79-27-6 | C2H2Br4 | 345.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-27-6
Molecular Formula C2H2Br4
Molecular Weight (g/mol) 345.65
MDL Number MFCD00000133
InChI Key QXSZNDIIPUOQMB-UHFFFAOYSA-N
Synonym acetylene tetrabromide, muthmann's liquid, ethane, 1,1,2,2-tetrabromo, tetrabromoacetylene, sym-tetrabromoethane, s-tetrabromoethane, 1,1,2,2-tetrabromaethan, 1,1,2,2-tetrabromoetano, tbe, unii-v00gkc584o
PubChem CID 6588
IUPAC Name 1,1,2,2-tetrabromoethane
SMILES BrC(Br)C(Br)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 1°C
Density 2.9600g/mL
Boiling Point 244°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6355 to 1.638
Linear Formula Br2CHCHBr2
Quantity 250 g
Beilstein 01, 94
Specific Gravity 2.96
Merck Index 15, 9330
Solubility Information Solubility in water: 0.63g/L (20°C). Other solubilities: soluble in acetone,benzene,miscible with ethanol,ether,chloroform,aniline,and glacial acetic acid
Formula Weight 345.64
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrabromoethane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if inhaled.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-191-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.