Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrachloroethane, 98.5%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
1L
2.5L
Packaging:
Glass bottle
79-34-5
C2H2Cl4
167.85
MFCD00000848
QPFMBZIOSGYJDE-UHFFFAOYSA-N
s-tetrachloroethane,acetylene tetrachloride,ethane, 1,1,2,2-tetrachloro,bonoform,cellon,tetrachlorethane,sym-tetrachloroethane,westron,1,1,2,2-tetrachlorethane,1,1,2,2-tetrachloraethan
6591
CHEBI:36026
1,1,2,2-tetrachloroethane
C(C(Cl)Cl)(Cl)Cl
79-34-5
C2H2Cl4
167.85
MFCD00000848
QPFMBZIOSGYJDE-UHFFFAOYSA-N
s-tetrachloroethane,acetylene tetrachloride,ethane, 1,1,2,2-tetrachloro,bonoform,cellon,tetrachlorethane,sym-tetrachloroethane,westron,1,1,2,2-tetrachlorethane,1,1,2,2-tetrachloraethan
6591
CHEBI:36026
1,1,2,2-tetrachloroethane
C(C(Cl)Cl)(Cl)Cl
Specific Gravity 1.58
Packaging Glass bottle
Refractive Index 1.4930 to 1.4950
Infrared Spectrum Authentic
Viscosity 1.7 mPa.s (28°C)
Color Colorless
Melting Point -43.0°C
Quantity 100mL
Density 1.5800g/mL
Boiling Point 147.0°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98.5%
Linear Formula Cl2CHCHCl2
Beilstein 01, 86
Merck Index 15, 9334
Solubility Information Solubility in water: 0.3 g/100mL (25°C). Other solubilities: miscible with methanol, ethanol, benzene, ether,, petroleum ether, carbon tetrachloride, chloroform,, carbon disulfide, dimethylformamide and oils, soluble in acetone
Formula Weight 167.85
Physical Form Liquid
Percent Purity 98.5%
Chemical Name or Material 1,1,2,2-Tetrachloroethane, 98.5%
Show More Show Less

GHS H Statement
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE

GHS Signal Word: Danger

EINECSNumber : 201-197-8

RTECSNumber : KI8575000

TSCA : TSCA