Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,3,3-Tetramethylurea 99%, ACROS Organics™

Click to view available options
Quantity:
100mL
250mL
1L
Packaging:
Glass bottle
632-22-4
C5H12N2O
116.16
MFCD00008319
AVQQQNCBBIEMEU-UHFFFAOYSA-N
tetramethylurea,temur,urea, tetramethyl,n,n,n',n'-tetramethylurea,tetramethyluree,tmu,urea, 1,1,3,3-tetramethyl,urea, n,n,n',n'-tetramethyl,tetramethyluree french,tetramethyl-urea
12437
CHEBI:84278
1,1,3,3-tetramethylurea
CN(C)C(=O)N(C)C
632-22-4
C5H12N2O
116.16
MFCD00008319
AVQQQNCBBIEMEU-UHFFFAOYSA-N
tetramethylurea,temur,urea, tetramethyl,n,n,n',n'-tetramethylurea,tetramethyluree,tmu,urea, 1,1,3,3-tetramethyl,urea, n,n,n',n'-tetramethyl,tetramethyluree french,tetramethyl-urea
12437
CHEBI:84278
1,1,3,3-tetramethylurea
CN(C)C(=O)N(C)C
Viscosity 1.5 mPa.s (20°C)
Melting Point -1.0°C
Refractive Index 1.4496 to 1.4516
Density 0.9879g/mL
Color Colorless to Yellow
Boiling Point 174.0°C to 178.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Water 0.5% max. (K.F.)
Specific Gravity 0.9879
Quantity 250mL
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2NCON(CH3)2
Beilstein 04, 74
Fieser 01,1146; 13,302; 15,305
Merck Index 15, 9378
Solubility Information Solubility in water: 116g/L. Other solubilities: miscible with all common organic solvents,,including petroleum ether
Formula Weight 116.16
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1,1,3,3-Tetramethylurea, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May damage the unborn child.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 211-173-9

RTECSNumber : YU2625000

TSCA : TSCA