Learn More
1,1,4,7,10,10-Hexamethyltriethylenetetramine, 97%
Description
1,1,4,7,10,10-Hexamethyltriethylenetetramine may be used as reagent in the synthesis of ideal linear random copolymers containing both vinyl polymer and polyester units in a single polymer chain.1 1,1,4,7,10,10-Hexamethyltriethylenetetramine complexed with CuBr constitutes catalytic complex, used in the copolymerization of poly[ε-caprolactone] with N,N-dimethylamino-2-ethyl methacrylate monomers by atom-transfer radical polymerization (ATRP).2 It may be used as catalyst in the aqueous surface-initiated-ATRP to grow poly(N,N-dimethylacrylamide) (PDMA).3
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,1,4,7,10,10-Hexamethyltriethylenetetramine may be used as reagent in the synthesis of ideal linear random copolymers containing both vinyl polymer and polyester units in a single polymer chain.1 1,1,4,7,10,10-Hexamethyltriethylenetetramine complexed with CuBr constitutes catalytic complex, used in the copolymerization of poly[ε-caprolactone] with N,N-dimethylamino-2-ethyl methacrylate monomers by atom-transfer radical polymerization (ATRP).2 It may be used as catalyst in the aqueous surface-initiated-ATRP to grow poly(N,N-dimethylacrylamide) (PDMA).3
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications
Specifications
| Density | 0.847 |
| Boiling Point | 130°C (11 mmHg) |
| Flash Point | 102°C (216°F) |
| Molecular Formula | C12H30N4 |
| Refractive Index | 1.456 |
| Quantity | 1 g |
| UN Number | UN3725 |
| Synonym | 1,1,4,7,10,10-hexamethyltriethylenetetramine, hexamethyltriethylenetetramine, 2,5,8,11-tetramethyl-2,5,8,11-tetraazadodecane, hmteta, 1,2-ethanediamine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethyl, n1,n1'-ethane-1,2-diyl bis n1,n2,n2-trimethylethane-1,2-diamine, n,n,n',n,n',n'-hexamethyltriethylenetetramine, triethylenetetramine, 1,1,4,7,10,10-hexamethyl, 1,1,4,7,10,10-hexamethyl-triethylenetetramine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethylethylenediamine |
| Solubility Information | Soluble in water. |
| InChI Key | DWFKOMDBEKIATP-UHFFFAOYSA-N |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POLYAMINES, LIQUID, CORROSIVE, N.O.S. |
| EINECSNumber | 221-382-7 |
| RTECSNumber | KH8587135 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.