Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,4,7,7-Pentamethyldiethylenetriamine, 98+%

Catalog No. p-3737147
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 3030-47-5 | C9H23N3 | 173.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3030-47-5
Molecular Formula C9H23N3
Molecular Weight (g/mol) 173.3
MDL Number MFCD00014876
InChI Key UKODFQOELJFMII-UHFFFAOYSA-N
Synonym pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine
PubChem CID 18196
ChEBI CHEBI:39475
IUPAC Name N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine
SMILES CN(C)CCN(C)CCN(C)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 18°C
Color White
Density 0.8290g/mL
Boiling Point 200°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4415 to 1.4435
Linear Formula (CH3)2NCH2CH2N(CH3)CH2CH2N(CH3)2
Quantity 25 g
Specific Gravity 0.829
Solubility Information Solubility in water: soluble
Viscosity 1.52 mPa.s (20°C)
Formula Weight 173.3
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 1,1,4,7,7-Pentamethyldiethylenetriamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 221-201-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.