Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Bis(4-aminophenyl)cyclohexane 98.0+%, TCI America™
SDP

Catalog No. B207625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B207625G 25 g
B20765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B207625G Supplier TCI America Supplier No. B207625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3282-99-3
Molecular Formula C18H22N2
Molecular Weight (g/mol) 266.39
MDL Number MFCD00035700
InChI Key DJCVCYUEICJZBD-UHFFFAOYSA-N
PubChem CID 768325
IUPAC Name 4-[4-(4-aminophenyl)cyclohexyl]aniline
SMILES NC1=CC=C(C=C1)C1CCC(CC1)C1=CC=C(N)C=C1

Specifications

Melting Point 110°C
Quantity 25 g
Synonym 4,4′-Cyclohexylidenedianiline
Formula Weight 266.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Bis(4-aminophenyl)cyclohexane
EINECSNumber (4)-0045
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.