Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis(diphenylphosphino)ferrocene 96.0+%, TCI America™
SDP

Catalog No. p-7119433
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 12150-46-8
Molecular Formula C34H28FeP2
Molecular Weight (g/mol) 554.39
MDL Number MFCD00001422
InChI Key KZPYGQFFRCFCPP-UHFFFAOYSA-N
IUPAC Name λ²-iron(2+) bis(1-(diphenylphosphanyl)cyclopenta-2,4-dien-1-ide)
SMILES [Fe].c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 182°C
Quantity 1 g
Synonym 1,1′C-Ferrocenebis(diphenylphosphine)
Formula Weight 554.39
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Bis(diphenylphosphino)ferrocene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.