Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Bis(hydroxymethyl)cyclopropane 96.0+%, TCI America™
SDP

Catalog No. B536525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B536525G 25 g
B53655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B536525G Supplier TCI America Supplier No. B536525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39590-81-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00464207
InChI Key YAINYZJQSQEGND-UHFFFAOYSA-N
Synonym 1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f
PubChem CID 4101437
IUPAC Name [1-(hydroxymethyl)cyclopropyl]methanol
SMILES C1CC1(CO)CO

Specifications

Color Yellow
Boiling Point 98°C
Quantity 25 g
Formula Weight 102.13
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1-Bis(hydroxymethyl)cyclopropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.