Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Carbonyldiimidazole, 97%

Catalog No. p-7022081
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 530-62-1 | C7H6N4O | 162.15 g/mol

Peptide coupling reagent1,1'-Carbonyldiimidazole acts as a coupling reagent and utilized for coupling of amino acids in order to prepare peptide in organic synthesis. It is also used in the preparation of beta-keto sulfones, sulfoxides and beta-enamino acid derivatives. It is used to convert alcohols and amines into carbamates, esters, and ureas. It is involved in the preparation of formylized imidazole by reaction with formic acid. Further, it is used in the synthesis of dipolar polyamides compounds. In addition to this, it is considered as an equivalent of phosgene and used to prepare asymmetric bis alkyl carbonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Peptide coupling reagent1,1′-Carbonyldiimidazole acts as a coupling reagent and utilized for coupling of amino acids in order to prepare peptide in organic synthesis. It is also used in the preparation of beta-keto sulfones, sulfoxides and beta-enamino acid derivatives. It is used to convert alcohols and amines into carbamates, esters, and ureas. It is involved in the preparation of formylized imidazole by reaction with formic acid. Further, it is used in the synthesis of dipolar polyamides compounds. In addition to this, it is considered as an equivalent of phosgene and used to prepare asymmetric bis alkyl carbonate.

Solubility
Soluble in dimethylformamide.

Notes
Store in a cool place. Incompatible with water, strong acids, strong oxidizing agents, strong bases and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-62-1
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.15
MDL Number MFCD00005286
InChI Key PFKFTWBEEFSNDU-UHFFFAOYSA-N
Synonym 1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole
PubChem CID 68263
SMILES O=C(N1C=CN=C1)N1C=CN=C1

Specifications

Melting Point 116°C to 120°C
pH 7.7
Assay Percent Range 97%
Quantity 10 g
UN Number UN3263
Beilstein 6826
Sensitivity Moisture sensitive
Merck Index 14,1819
Solubility Information Soluble in dimethylformamide.
IUPAC Name di(imidazol-1-yl)methanone
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 1,1'-Carbonyldiimidazole
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber 208-488-9
TSCA Yes
storageNote1 Moisture sensitive
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.