Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Carbonyldiimidazole, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
500g
Packaging:
Glass bottle
530-62-1
C7H6N4O
162.15
MFCD00005286
PFKFTWBEEFSNDU-UHFFFAOYSA-N
1,1'-carbonyldiimidazole,n,n'-carbonyldiimidazole,carbonyldiimidazole,di 1h-imidazol-1-yl methanone,n,n-carbonyldiimidazole,1h-imidazole, 1,1'-carbonylbis,1,1'-carbonylbis-1h-imidazole,carbonyl diimidazole,diimidazol-1-yl ketone,1,1-carbonyldiimidazole
68263
di(imidazol-1-yl)methanone
C1=CN(C=N1)C(=O)N2C=CN=C2
530-62-1
C7H6N4O
162.15
MFCD00005286
PFKFTWBEEFSNDU-UHFFFAOYSA-N
1,1'-carbonyldiimidazole,n,n'-carbonyldiimidazole,carbonyldiimidazole,di 1h-imidazol-1-yl methanone,n,n-carbonyldiimidazole,1h-imidazole, 1,1'-carbonylbis,1,1'-carbonylbis-1h-imidazole,carbonyl diimidazole,diimidazol-1-yl ketone,1,1-carbonyldiimidazole
68263
di(imidazol-1-yl)methanone
C1=CN(C=N1)C(=O)N2C=CN=C2
Coupling reagent
Melting Point 118.0°C to 120.0°C
Packaging Glass bottle
Quantity 500g
Color White to Cream
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 23, V,4, 245
Fieser 01,114; 02,61; 05,97; 06,97; 08,77; 09,96; 12,106; 13,66; 16,64
Merck Index 15, 1820
Solubility Information Solubility in water: decomposes. Other solubilities: moderately soluble in thf, soluble in ethanol
Formula Weight 162.15
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 1,1′-Carbonyldiimidazole
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger