Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Cyclohexanediacetic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass Bottle
4355-11-7
C10H16O4
200.23
MFCD00001519
YQPCHPBGAALCRT-UHFFFAOYSA-N
1,1-cyclohexanediacetic acid,1,1-cyclohexanediaceticacid,cyclohexanediacetic acid,2-1-carboxymethyl cyclohexyl acetic acid,1,1-cyclohexane diacetic acid,1-carboxymethyl cyclohexyl acetic acid,2,2'-cyclohexane-1,1-diyl diacetic acid,2-carboxymethyl cyclohexyl acetic acid,gabapentin impurity 2,pubchem13645
78061
2-[1-(carboxymethyl)cyclohexyl]acetic acid
C1CCC(CC1)(CC(=O)O)CC(=O)O
4355-11-7
C10H16O4
200.23
MFCD00001519
YQPCHPBGAALCRT-UHFFFAOYSA-N
1,1-cyclohexanediacetic acid,1,1-cyclohexanediaceticacid,cyclohexanediacetic acid,2-1-carboxymethyl cyclohexyl acetic acid,1,1-cyclohexane diacetic acid,1-carboxymethyl cyclohexyl acetic acid,2,2'-cyclohexane-1,1-diyl diacetic acid,2-carboxymethyl cyclohexyl acetic acid,gabapentin impurity 2,pubchem13645
78061
2-[1-(carboxymethyl)cyclohexyl]acetic acid
C1CCC(CC1)(CC(=O)O)CC(=O)O
Melting Point 181°C to 185°C
Water 1% max.
Packaging Glass Bottle
Color White
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H10(CH2CO2H)2
Formula Weight 200.23
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 1, 1-Cyclohexanediacetic acid, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 224-427-9

TSCA : TSCA