Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dichloroethylene, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187374
Click to view available options
Quantity:
1 mL
5 mL

1,1-Dichloroethylenes are a group of volatile organic compounds (VOCs), which are characterized by high vapor pressure and low water solubility.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-35-4
Ignition Point 968°F
Solution Contains ˜0.02% 4-methoxyphenol as stabilizer
Vapor Pressure 9.68 psi (20°C)
Vapor Density 3.46 (vs. air)
Density 1.213 g/mL (at 20°C)
Flash Point -19°C - closed cup
Linear Formula CH2=CCl2
MDL Number MFCD00011653
Melting Point -122°C
Boiling Point 30°C to 32°C
Synonym Vinylidene chloride
Type Analytical Standard
Quantity 1 mL
Formula Weight 96.94
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.