Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Diethyl-2,2'-cyanine iodide, 99%

Catalog No. ac407255000
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle

CAS: 977-96-8 | C23H23IN2 | 454.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 977-96-8
Molecular Formula C23H23IN2
Molecular Weight (g/mol) 454.34
MDL Number MFCD00011971
InChI Key GMYRVMSXMHEDTL-UHFFFAOYSA-M
Synonym decynium 22
PubChem CID 71299759
ChEBI CHEBI:37993
IUPAC Name (2Z)-1-ethyl-2-[(1-ethylquinolin-1-ium-2-yl)methylidene]quinoline;iodide
SMILES CCN1C(=CC2=[N+](C3=CC=CC=C3C=C2)CC)C=CC4=CC=CC=C41.[I-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 273°C
Color Red to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 500 mg
Beilstein 23, II, 267
Formula Weight 454.34
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Diethyl-2,2′-cyanine iodide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
EINECSNumber 213-556-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.