Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-diethylferrocene 98%, Thermo Scientific™

Catalog No. ac381100010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1273-97-8
MDL Number MFCD01862455

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown-Red to Orange
Density 1.1800g/mL
Boiling Point 284.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C14H18Fe
Refractive Index 1.5770 to 1.5800
Quantity 1 g
Specific Gravity 1.18
Solubility Information Solubility in water: soluble.
Formula Weight 242.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1′-Diethylferrocene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.