Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dimethyl-4-phenylpiperazinium Iodide 99%, Thermo Scientific™

Catalog No. AC116620010
Click to view available options
Quantity:
1 g
10 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-77-3
Molecular Formula C12H19IN2
Molecular Weight (g/mol) 318.20
MDL Number MFCD00011976
InChI Key XFZJGFIKQCCLGK-UHFFFAOYSA-M
Synonym 1,1-dimethyl-4-phenylpiperazinium iodide, dmpp, dmpp iodide, dimethylphenylpiperazinium iodide, 1,1-dimethyl-4-phenylpiperazin-1-ium iodide, 1,1-dimethyl-4-phenylpiperazine iodide, n,n-dimethyl-n'-phenylpiperazinium iodide, dimethyl-4-phenylpiperazium iodide, 1,1-dimethyl-4-phenyl-piperazinium iodide, unii-x95h7drx99
PubChem CID 5911
ChEBI CHEBI:4290
IUPAC Name 1,1-dimethyl-4-phenylpiperazin-1-ium iodide
SMILES [I-].C[N+]1(C)CCN(CC1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 234°C to 238°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 1 g
Formula Weight 318.19
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1, 1-Dimethyl-4-phenylpiperazinium iodide, 99%
Show More Show Less
EINECSNumber 200-213-0
RTECSNumber TM3385000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.