Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dimethylpropargylamine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2978-58-7
C5H9N
83.13
MFCD00008052
VUGCBIWQHSRQBZ-UHFFFAOYSA-N
1,1-dimethylpropargylamine,3-amino-3-methyl-1-butyne,3-butyn-2-amine, 2-methyl,1,1-dimethylprop-2-ynylamine,2-methyl-3-butyn-2-amine,1,1-dimethyl-prop-2-ynylamine,1,1-dimethylpropynylamine,1,1-dimethyl-2-propynylamine,1,1-dimethylprop-3-ynylamine,3-amino-3-methylbutyne
76319
2-methylbut-3-yn-2-amine
CC(C)(C#C)N
2978-58-7
C5H9N
83.13
MFCD00008052
VUGCBIWQHSRQBZ-UHFFFAOYSA-N
1,1-dimethylpropargylamine,3-amino-3-methyl-1-butyne,3-butyn-2-amine, 2-methyl,1,1-dimethylprop-2-ynylamine,2-methyl-3-butyn-2-amine,1,1-dimethyl-prop-2-ynylamine,1,1-dimethylpropynylamine,1,1-dimethyl-2-propynylamine,1,1-dimethylprop-3-ynylamine,3-amino-3-methylbutyne
76319
2-methylbut-3-yn-2-amine
CC(C)(C#C)N
Specific Gravity 0.79
Density 0.7900g/mL
Boiling Point 79.0°C to 80.0°C
Flash Point 2°C
Refractive Index 1.4220 to 1.4250
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula HC≡CC(CH3)2NH2
Formula Weight 83.13
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 1, 1-Dimethylpropargylamine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 221-029-7