Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dimethylsemicarbazide 96.0+%, TCI America™
SDP

Catalog No. D385425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D385425G 25 g
D38545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D385425G Supplier TCI America Supplier No. D385425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22718-49-6
Molecular Formula C3H9N3O
Molecular Weight (g/mol) 103.125
InChI Key SVZFWZRCMLRFOM-UHFFFAOYSA-N
PubChem CID 14799320
IUPAC Name dimethylaminourea
SMILES CN(C)NC(=O)N

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 25 g
Formula Weight 103.13
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Dimethylsemicarbazide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.