Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,1-Diphenyl-2-picrylhydrazine 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
Specifications
Specifications
| CAS | 1707-75-1 |
| Molecular Formula | C18H13N5O6 |
| Quantity | 25 g |
| MDL Number | MFCD00007229 |
| UN Number | 1325 |
| InChI Key | WCBPJVKVIMMEQC-UHFFFAOYSA-N |
| SMILES | C1=CC=C(C=C1)N(C2=CC=CC=C2)NC3=C(C=C(C=C3[N+](=O)[O-])[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | 1,1-diphenyl-2-(2,4,6-trinitrophenyl)hydrazine |
| Molecular Weight (g/mol) | 395.331 |
| PubChem CID | 74358 |
| Show More |
Safety and Handling
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction