Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Diphenylethylene oxide, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
882-59-7
C14H12O
196.25
MFCD00040725
PRLJMHVNHLTQJJ-UHFFFAOYSA-N
1,1-diphenylethylene oxide,oxirane, 2,2-diphenyl,diphenyloxirane,2,2-diphenyl-oxirane,2,2-diphenyl oxirane,#,1,1-diphenyl-ethylenoxide,1,1-diphenyl-ethylene oxide,1,1-diphenyl-1,2-epoxyethane
93564
2,2-diphenyloxirane
C1C(O1)(C2=CC=CC=C2)C3=CC=CC=C3
882-59-7
C14H12O
196.25
MFCD00040725
PRLJMHVNHLTQJJ-UHFFFAOYSA-N
1,1-diphenylethylene oxide,oxirane, 2,2-diphenyl,diphenyloxirane,2,2-diphenyl-oxirane,2,2-diphenyl oxirane,#,1,1-diphenyl-ethylenoxide,1,1-diphenyl-ethylene oxide,1,1-diphenyl-1,2-epoxyethane
93564
2,2-diphenyloxirane
C1C(O1)(C2=CC=CC=C2)C3=CC=CC=C3
Melting Point 54°C to 56°C
Infrared Spectrum Authentic
Quantity 1g
Color White
Packaging Glass bottle
Formula Weight 196.25
Physical Form Crystals
Percent Purity 96%
Chemical Name or Material 1, 1-Diphenylethylene oxide

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-929-