Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Naphthyl)-2-thiourea 97%, Thermo Scientific™

Catalog No. AC415430250
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-88-4
Molecular Formula C11H10N2S
Molecular Weight (g/mol) 202.275
MDL Number MFCD00041824
InChI Key PIVQQUNOTICCSA-UHFFFAOYSA-N
Synonym 1-1-naphthyl-2-thiourea, antu, 1-naphthylthiourea, dirax, anturat, rattrack, smeesana, alrato, bantu, kripid
PubChem CID 736366
ChEBI CHEBI:82256
IUPAC Name naphthalen-1-ylthiourea
SMILES C1=CC=C2C(=C1)C=CC=C2NC(=S)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Beige-Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 25 g
Merck Index 15, 708
Formula Weight 202.28
Percent Purity ≥96.0%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1-(1-Naphthyl)-2-thiourea, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Fatal if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.