Alfa Aesar™ 1-(1-Naphthyl)-2-(trimethylsilyl)acetylene, 97%

Manufacturer: Alfa Aesar™ H5595303

Quantity 1g

 View more versions of this product

Catalog No. AAH5595303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 104784-51-2
    Formula Weight 224.38
    MDL Number MFCD04039886
    Synonym Trimethyl[(1-naphthyl)ethynyl]silane
    Chemical Name or Material 1-(1-Naphthyl)-2-(trimethylsilyl)acetylene

    Specifications

    Formula Weight 224.38
    Melting Point 41° to 43°C
    Boiling Point 292° to 293°C
    Refractive Index 1.5950
    Physical Form Solid
    Density 0.968g/mL
    MDL Number MFCD04039886
    Quantity 1g
    CAS 104784-51-2
    Synonym Trimethyl[(1-naphthyl)ethynyl]silane
    Chemical Name or Material 1-(1-Naphthyl)-2-(trimethylsilyl)acetylene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit