Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Bis(acryloyloxy)decane (stabilized with MEHQ) 90.0+%, TCI America™
SDP

Catalog No. B2937100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2937100G 100 g
B293725G 25 g
B2937500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B2937100G Supplier TCI America Supplier No. B2937100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13048-34-5
Molecular Formula C16H26O4
Molecular Weight (g/mol) 282.38
MDL Number MFCD00080453
InChI Key RHNJVKIVSXGYBD-UHFFFAOYSA-N
Synonym 1,10-Decanediol Diacrylate, Decamethylene Glycol Diacrylate
PubChem CID 83079
IUPAC Name 10-(prop-2-enoyloxy)decyl prop-2-enoate
SMILES C=CC(=O)OCCCCCCCCCCOC(=O)C=C

Specifications

Color Yellow
Boiling Point 181°C
Quantity 100 g
Formula Weight 282.38
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,10-Bis(acryloyloxy)decane (stabilized with MEHQ)
EINECSNumber (2)-0958&(2)-1008
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.