Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Diamino-4,7-dioxadecane, 95%, Thermo Scientific™

Catalog No. AC371610050
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2997-01-5
Molecular Formula C8H22N2O2
Molecular Weight (g/mol) 178.28
MDL Number MFCD00059938
InChI Key POTQBGGWSWSMCX-UHFFFAOYSA-P

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.9837g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
Linear Formula NH2(CH2)3O(CH2)2O(CH2)3NH2
Quantity 5 g
Synonym diethylaminoacetonitrile, 2-diethylamino acetonitrile, acetonitrile, diethylamino, n,n-diethylaminoacetonitrile, nitril kiseliny diethylaminooctove, diethylamino acetonitrile, glycinonitrile, n,n-diethyl, unii-468yw0h2fa, acetonitrile, 2-diethylamino, nitril kiseliny diethylaminooctove czech
SMILES [NH3+]CCCOCCOCCC[NH3+]
IUPAC Name 3-[2-(3-azaniumylpropoxy)ethoxy]propan-1-aminium
PubChem CID 44983
Formula Weight 176.26
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material 1, 10-Diamino-4, 7-dioxadecane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.