Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Diaminodecane 98.0+%, TCI America™
SDP

Catalog No. D0085100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0085100G 100 g
D008525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0085100G Supplier TCI America Supplier No. D0085100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 646-25-3
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.316
MDL Number MFCD00008151
InChI Key YQLZOAVZWJBZSY-UHFFFAOYSA-N
Synonym 1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine
PubChem CID 1317
IUPAC Name decane-1,10-diamine
SMILES C(CCCCCN)CCCCN

Specifications

Melting Point 61°C
Color White-Yellow
Boiling Point 140°C
Quantity 100 g
UN Number 3259
Formula Weight 172.32
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,10-Diaminodecane
EINECSNumber (2)-3742
RTECSNumber HD7175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.