Learn More
1,10-Diaza-18-crown-6, 96%
Description
1,10-Diaza-18-crown-6 are starting materials to produce macrocyclic polyamine metal-complexing agents. It is used as a ligand in chemistry for the applications of diagnostic imaging technique to identify medical condition or disease and for the development of medication as a therapeutic agent and pharmaceutic aid. Also used as an ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Specifications
Specifications
Quantity | 1 g |
Beilstein | 609764 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 262.35 |
Percent Purity | 96% |
Chemical Name or Material | 1,10-Diaza-18-crown-6 |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
EINECSNumber | 245-965-0 |
RTECSNumber | HM1650000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.