Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Phenanthroline, 0.1% (w/v) Alcoholic, Ricca Chemical
SDP

Catalog No. 5520516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5520-516 500 mL Amber Glass Bottle
R55205001C 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5520-516 Supplier Ricca Chemical Company Supplier No. 5520.516
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5144-89-8 , 67-56-1
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
IUPAC Name 1,10-phenanthroline hydrate
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Color Colorless to Green
CAS Min % 98.0, 0.12
CAS Max % 100.0, 0.13
MDL Number MFCD00149973
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
PubChem CID 21226
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.1% (w/v) Alcoholic
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material 1, 10-Phenanthroline
Show More Show Less
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.