Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Phenanthroline-5,6-dione, 98%

Catalog No. AAH5574703
Click to view available options
Quantity:
250 mg
1 g
5 g

Catalog No. AAH5574703

Supplier: Thermo Scientific Chemicals H5574703

Only null left
Add to Cart

CAS: 27318-90-7 | C12H6N2O2 | 210.19 g/mol

1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.

Solubility
Insoluble in water.

Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27318-90-7
Molecular Formula C12H6N2O2
Molecular Weight (g/mol) 210.19
MDL Number MFCD00014473
InChI Key KCALAFIVPCAXJI-UHFFFAOYSA-N
Synonym 1,10 phenanthroline-5,6-dione, 1 10-phenanthroline-5 6-dione 97, pyridino 3,2-h quinoline-5,6-dione, phen-5,6-dione, pubchem16755, acmc-209gwd, d0jh7c, phox31, ksc204s8l, 1,2-dione-based compound, 17
PubChem CID 72810
IUPAC Name 1,10-phenanthroline-5,6-dione
SMILES O=C1C(=O)C2=CC=CN=C2C2=NC=CC=C12

Specifications

Melting Point 260°C (decomposition)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 210.19
Percent Purity 98%
Chemical Name or Material 1,10-Phenanthroline-5,6-dione
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.