Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Phenanthroline monohydrate, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
5144-89-8,66-71-7
C12H8N2·H2O
198.23
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate
21226
1,10-phenanthroline;hydrate
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O
5144-89-8,66-71-7
C12H8N2·H2O
198.23
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate
21226
1,10-phenanthroline;hydrate
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O
Indicator
Melting Point 97.0°C to 101.0°C
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Color Cream-White
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Beilstein 23,227
Merck Index 15,7328
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, acetone and benzene
Formula Weight 198.23
Physical Form Needle-Like Crystalline Powder
Percent Purity ≥99%
Grade Pure
Chemical Name or Material 1, 10-Phenanthroline monohydrate
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger