Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1, 10-Phenanthroline Monohydrate, ACS Reagent, Reagents

Catalog No. 01337143
Change view
Click to view available options
Quantity:
5 g
25 g
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-143 25 g
01-337-145 100 mL
01-337-142 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-143 Supplier Reagents Holdings Llc Supplier No. C226770025C2
Add to Cart
Add to Cart

1, 10-Phenanthroline Monohydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5144-89-8
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
Synonym o-Phenanthroline Monohydrate
IUPAC Name 1,10-phenanthroline hydrate
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Color White to Off-White
Packaging Glass Bottle
Quantity 25 g
Specific Gravity 1.1
Grade ACS
Physical Form Solid
Chemical Name or Material 1, 10-Phenanthroline Monohydrate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.