Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Undecadiene 90.0+%, TCI America™
SDP

Catalog No. U00241ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00241ML 1 mL
U00245ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00241ML Supplier TCI America Supplier No. U00241ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13688-67-0
Molecular Formula C11H20
Molecular Weight (g/mol) 152.281
MDL Number MFCD00059203
InChI Key VOSLXTGMYNYCPW-UHFFFAOYSA-N
PubChem CID 139543
IUPAC Name undeca-1,10-diene
SMILES C=CCCCCCCCC=C

Specifications

Color Yellow
Boiling Point 187°C
Quantity 1 mL
UN Number 3295
Formula Weight 152.28
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,10-Undecadiene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.