Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,11-Dioxa[11]paracyclophane 97.0+%, TCI America™
SDP

Catalog No. D42981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42981G 1 g
D4298250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42981G Supplier TCI America Supplier No. D42981G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6571-51-3
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.34
MDL Number MFCD28100826
InChI Key LTROLYACVISZRU-UHFFFAOYSA-N
PubChem CID 91972135
IUPAC Name 2,12-dioxabicyclo[11.2.2]heptadeca-1(15),13,16-triene
SMILES C1CCCCOC2=CC=C(OCCCC1)C=C2

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 1 g
Formula Weight 234.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,11-Dioxa[11]paracyclophane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.