Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,12-Diaminododecane 98.0+%, TCI America™
SDP

Catalog No. D0091100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0091100G 100 g
D009125G 25 g
D0091500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0091100G Supplier TCI America Supplier No. D0091100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2783-17-7
Molecular Formula C12H28N2
Molecular Weight (g/mol) 200.37
MDL Number MFCD00008155
InChI Key QFTYSVGGYOXFRQ-UHFFFAOYSA-N
Synonym 1,12-diaminododecane, 1,12-dodecanediamine, dodecamethylenediamine, dodecyldiamine, 1,12-dodecamethylenediamine, 1,12-diamindodecane, 1,12'-dodecylenediamine, 1,12'-dodecamethylenediamine, unii-j3lm80w9nt, 1,12-dodecylenediamine
PubChem CID 17727
ChEBI CHEBI:49385
IUPAC Name dodecane-1,12-diamine
SMILES C(CCCCCCN)CCCCCN

Specifications

Melting Point 70°C
Color White
Boiling Point 145°C
Quantity 100 g
Formula Weight 200.37
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,12-Diaminododecane
EINECSNumber (2)-2370
RTECSNumber JR2200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.