Learn More
1,12-Dodecanediol, 99%
Description
1,12-Dodecanediol is used in the polyester polyols for polyurethanes, coatings and inks, adhesives, lastomers, polyesters and co polyesters, polymer cross linkers, pharmaceutical intermediates, fragrances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,12-Dodecanediol is used in the polyester polyols for polyurethanes, coatings and inks, adhesives, lastomers, polyesters and co polyesters, polymer cross linkers, pharmaceutical intermediates, fragrances.
Solubility
Soluble in alcohol and warm ether. Insoluble in water and petroleum ether.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 81°C to 84°C |
| Boiling Point | 188°C to 190°C (12 mmHg) |
| Quantity | 25 g |
| Beilstein | 1742760 |
| Solubility Information | Soluble in alcohol and warm ether. Insoluble in water and petroleum ether. |
| Formula Weight | 202.34 |
| Percent Purity | 99% |
| Chemical Name or Material | 1,12-Dodecanediol |
Safety and Handling
| EINECSNumber | 227-133-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.