Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,12-Dodecanediol, 98%

Catalog No. ac117610250
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle

CAS: 5675-51-4 | C12H26O2 | 202.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5675-51-4
Molecular Formula C12H26O2
Molecular Weight (g/mol) 202.34
MDL Number MFCD00004755
InChI Key GHLKSLMMWAKNBM-UHFFFAOYSA-N
Synonym 1,12-dodecanediol, dodecamethylene glycol, chembl17440, 1,12-dihydroxy dodecane, 1,12-dodecandiol, 1,12-dodecane diol, acmc-1ambo, 1,12-dihydroxydodecane, ksc270c5h
PubChem CID 79758
IUPAC Name dodecane-1,12-diol
SMILES OCCCCCCCCCCCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 79.0°C to 81.0°C
Color Brown to Orange
Boiling Point 324.0°C
Flash Point 176°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HO(CH2)12OH
Quantity 25 g
Beilstein 01,II,562
Solubility Information Solubility in water: <0.1% in water.
Viscosity 16 mPa.s (85°C)
Formula Weight 202.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 12-Dodecanediol, 99%
Show More Show Less
EINECSNumber 227-133-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.