Learn More
Restek™ 1/16 in. Capillary Inlet Adaptor Fitting Kit
SDP
Catalog No. 06611273 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent Capillary Injectors
Catalog No. For Use With (Equipment)
06-611-273 Agilent Capillary Injectors
1 options
Catalog No. 06-611-273 Supplier Restek™ Supplier No. 27184
Split/Splitless Fitting Accepts Standard Capillary Ferrules
  • Capillary columns are easier to install because the nut protrudes farther from the insulated injection port chamber
  • Same column insertion depth as the original manufacturer's equipment
  • Includes Adaptor Fitting, Capillary Nut, Gold Plated Dual Vespel Ring Inlet Seal (0.8 mm I.D.), Ferrule (0.4 mm I.D.)
Specifications
Type Capillary Inlet Adaptor Fitting Kit
Includes Adaptor Fitting, Capillary Nut, Gold Plated Dual Vespel Ring Inlet Seal (0.8 mm I.D.), Ferrule (0.4 mm I.D.)
Size (English) 1/16 in.
For Use With (Application) Single Column Installation
For Use With (Equipment) Agilent Capillary Injectors