Learn More
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane (1:1), Pd 13%, Thermo Scientific Chemicals
Description
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II) is used for palladium-catalyzed coupling reactions. It serves as a catalyst and also employed in carbonylation reactions. Further, it is used in cross-coupling reaction and Suzuki reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 275°C to 280°C |
Quantity | 1 g |
Synonym | Dichloro[1, 1′-bis(diphenylphosphino)ferrocene]palladium(II) dichloromethane complex |
Solubility Information | Insoluble in water. |
Formula Weight | 816.64 |
Physical Form | Powder |
Chemical Name or Material | [1, 1′-Bis(diphenylphosphino)ferrocene]palladium(II) chloride |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H335 |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.