Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-1H-Triazole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
288-36-8
C2H3N3
69.07
QWENRTYMTSOGBR-UHFFFAOYSA-N
1h-1,2,3-triazole,1,2,3-triazole,2h-1,2,3-triazole,1,2,3-1h-triazole,triazole,triazol,osotriazole,v-triazole,1,2,3-triazol,1,2,3 triazole
67516
CHEBI:35565
2H-triazole
C1=NNN=C1
288-36-8
C2H3N3
69.07
QWENRTYMTSOGBR-UHFFFAOYSA-N
1h-1,2,3-triazole,1,2,3-triazole,2h-1,2,3-triazole,1,2,3-1h-triazole,triazole,triazol,osotriazole,v-triazole,1,2,3-triazol,1,2,3 triazole
67516
CHEBI:35565
2H-triazole
C1=NNN=C1
Refractive Index 1.4930 to 1.4960
Quantity 1g
Flash Point 107°C
Packaging Glass bottle
Specific Gravity 1.192
Infrared Spectrum Authentic
Color Colorless to Yellow
Melting Point 23.0°C to 25.0°C
Boiling Point 203.0°C (752.0 mmHg)
Density 1.1920g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,diethyl ether,insoluble in n-pentane
Formula Weight 69.07
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1, 2, 3-1H-Triazole
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning