Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetra-O-acetyl-alpha-L-fucopyranose 98.0+%, TCI America™
SDP

Catalog No. T22071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22071G 1 g
T22075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22071G Supplier TCI America Supplier No. T22071G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64913-16-2
Molecular Formula C14H20O9
Molecular Weight (g/mol) 332.305
MDL Number MFCD00069791
InChI Key QZQMGQQOGJIDKJ-CSHNMWLISA-N
Synonym alpha-L-Fucopyranose 1,2,3,4-Tetraacetate
PubChem CID 11088873
IUPAC Name [(2S,3R,4R,5S,6S)-4,5,6-triacetyloxy-2-methyloxan-3-yl] acetate
SMILES CC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 94°C
Color White
Quantity 1 g
Formula Weight 332.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Tetra-O-acetyl-alpha-L-fucopyranose
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.