Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrachlorobenzene 96.0+%, TCI America™
SDP

Catalog No. T063125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T063125G 25 g
T0631500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T063125G Supplier TCI America Supplier No. T063125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 634-66-2
Molecular Formula C6H2Cl4
Molecular Weight (g/mol) 215.88
MDL Number MFCD00000538
InChI Key GBDZXPJXOMHESU-UHFFFAOYSA-N
Synonym benzene, 1,2,3,4-tetrachloro, tetrachlorobenzene, caswell no. 825, unii-mh0uy3v1ke, ccris 5935, 1,2,3,4-benzene tetrachloride, mh0uy3v1ke, epa pesticide chemical code 061101, 1,2,3,4-tetrachlorbenzol, 1,2,3,4-tetrachloro-benzene
PubChem CID 12463
ChEBI CHEBI:18855
IUPAC Name 1,2,3,4-tetrachlorobenzene
SMILES ClC1=CC=C(Cl)C(Cl)=C1Cl

Specifications

Melting Point 45°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 3077
Boiling Point 254°C
Formula Weight 215.88
Percent Purity ≥96.0% (GC)
Chemical Name or Material 1,2,3,4-Tetrachlorobenzene
EINECSNumber (3)-0076
RTECSNumber DB9440000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.