Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydro-1-naphthylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2217-40-5
C10H13N
147.22
MFCD00001740
JRZGPXSSNPTNMA-UHFFFAOYSA-N
1,2,3,4-tetrahydro-1-naphthylamine,1-aminotetralin,aminotetralin,1-amino-1,2,3,4-tetrahydro-naphthalene,aminotetralin czech,tetrahydro-beta-naphthylamine,1-amino-1,2,3,4-tetrahydronaphthalene,1-naphthalenamine, 1,2,3,4-tetrahydro,1,2,3,4-tetrahydronaphthylamine,beta-1,2,3,4-tetrahydronaphthylamine
18066
1,2,3,4-tetrahydronaphthalen-1-amine
C1CC(C2=CC=CC=C2C1)N
2217-40-5
C10H13N
147.22
MFCD00001740
JRZGPXSSNPTNMA-UHFFFAOYSA-N
1,2,3,4-tetrahydro-1-naphthylamine,1-aminotetralin,aminotetralin,1-amino-1,2,3,4-tetrahydro-naphthalene,aminotetralin czech,tetrahydro-beta-naphthylamine,1-amino-1,2,3,4-tetrahydronaphthalene,1-naphthalenamine, 1,2,3,4-tetrahydro,1,2,3,4-tetrahydronaphthylamine,beta-1,2,3,4-tetrahydronaphthylamine
18066
1,2,3,4-tetrahydronaphthalen-1-amine
C1CC(C2=CC=CC=C2C1)N
Refractive Index 1.5625 to 1.5645
Specific Gravity 1.02
Quantity 5g
Infrared Spectrum Authentic
Boiling Point 246.0°C to 247.0°C (714.0mmHg)
Color Colorless to Yellow
Flash Point >110°C
Packaging Glass bottle
Density 1.0200g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Beilstein 12,12
Formula Weight 147.22
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1,2,3,4-Tetrahydro-1-naphthylamine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 218-712-7