Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydro-2-naphthol, 97%

Catalog No. ac271880010
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 530-91-6 | C10H12O | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-91-6
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
MDL Number MFCD00045575
InChI Key JWQYZECMEPOAPF-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-2-naphthol, 2-hydroxytetralin, 2-tetralinol, 2-hydroxytetraline, beta-tetralol, tetrahydronaphthol-2, 2-naphthalenol, 1,2,3,4-tetrahydro, .beta.-tetralol, ac-tetrahydro-beta-naphthol, 2-naphthol, 1,2,3,4-tetrahydro
PubChem CID 10747
IUPAC Name 1,2,3,4-tetrahydronaphthalen-2-ol
SMILES C1CC2=CC=CC=C2CC1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 15.5°C
Color Brown to Yellow
Boiling Point 140°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5620 to 1.564
Quantity 1 g
Merck Index 15, 9369
Formula Weight 148.2
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material 1,2,3,4-Tetrahydro-2-naphthol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-497-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.