Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydronaphthalene, 98+%, pure

Catalog No. AC146730025
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum

CAS: 119-64-2 | C10H12 | 132.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-64-2
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00001733
InChI Key CXWXQJXEFPUFDZ-UHFFFAOYSA-N
Synonym tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene
PubChem CID 8404
ChEBI CHEBI:35008
IUPAC Name 1,2,3,4-tetrahydronaphthalene
SMILES C1CCC2=CC=CC=C2C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -35.0°C
Density 0.9730g/mL
Boiling Point 207.0°C
Flash Point 77°C
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5400 to 1.5420
Linear Formula C10H12
Quantity 2.5 L
Beilstein 05, IV, 1388
Fieser 01,1142
Specific Gravity 0.973
Merck Index 15, 9368
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents
Viscosity 2.2 mPa.s (20°C)
Formula Weight 132.2
Percent Purity 98+%
Grade Pure
Chemical Name or Material 1,2,3,4-Tetrahydronaphthalene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May form explosive peroxides.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
EINECSNumber 204-340-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.