Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydroquinoline, 98%

Catalog No. AAB2275036
Click to view available options
Quantity:
100 g
500 g

CAS: 635-46-1 | C9H11N | 133.19 g/mol

1,2,3,4-Tetrahydroquinoline is used as an antioxidant and corrosion inhibitor. It is an active component in various dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydroquinoline is used as an antioxidant and corrosion inhibitor. It is an active component in various dyes.

Solubility
Miscible with alcohol, ether, carbon disulfide and many organic solvents. Slightly miscible with water.

Notes
Hygroscopic. Incomaptible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-46-1
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00006693
InChI Key LBUJPTNKIBCYBY-UHFFFAOYSA-N
Synonym tetrahydroquinoline, 1,2,3,4-tetrahydro-quinoline, kusol, quinoline, 1,2,3,4-tetrahydro, unii-ccr50n1z9g, 1,2,3,4-tetrahydrochinoline, 1,2,3,4-tetrahydro quinoline, benzopiperidine, ccr50n1z9g, 1,2,3,4,-tetrahydroquinoline
PubChem CID 69460
ChEBI CHEBI:213323
IUPAC Name 1,2,3,4-tetrahydroquinoline
SMILES C1CNC2=CC=CC=C2C1

Specifications

Melting Point ∼14°C
Density 1.059
Boiling Point 248°C to 250°C
Flash Point 100°C (212°F)
Refractive Index 1.595
Quantity 500 g
Beilstein 116149
Solubility Information Miscible with alcohol,ether,carbon disulfide and many organic solvents. Slightly miscible with water.
Formula Weight 133.19
Percent Purity 99%
Chemical Name or Material 1,2,3,4-Tetrahydroquinoline
Show More Show Less
Hazard Category H315-H319-H335-H350
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 211-237-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.