Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,5-Tetrachlorobenzene 98.0+%, TCI America™
SDP

Catalog No. T087025G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 634-90-2
Molecular Formula C6H2Cl4
Molecular Weight (g/mol) 215.88
MDL Number MFCD00000543
InChI Key QZYNWJQFTJXIRN-UHFFFAOYSA-N
Synonym benzene, 1,2,3,5-tetrachloro, unii-i27n186cin, ccris 5936, acmc-209nfx, dsstox_cid_6089, 1,3,5-tetrachlorobenzene, 1,2,3,5-tetrachlorbenzol, dsstox_rid_78013, dsstox_gsid_26089, ksc454c5l
PubChem CID 12468
ChEBI CHEBI:36696
IUPAC Name 1,2,3,5-tetrachlorobenzene
SMILES ClC1=CC(Cl)=C(Cl)C(Cl)=C1

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 246°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 215.88
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,2,3,5-Tetrachlorobenzene
EINECSNumber (3)-0076
RTECSNumber DB9445000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.