Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Propanetricarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. P0490100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0490100G 100 g
P049025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0490100G Supplier TCI America Supplier No. P0490100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99-14-9
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00002723
InChI Key KQTIIICEAUMSDG-UHFFFAOYSA-N
Synonym tricarballylic acid, 1,2,3-propanetricarboxylic acid, carballylic acid, tricarballylate, beta-carboxyglutaric acid, propane 1,2,3-tricarboxylic acid, unii-ra5qh2j020, 3-carboxyglutaric acid, .beta.-carboxyglutaric acid, 1,2,3-propanetricarboxylicacid
PubChem CID 14925
ChEBI CHEBI:45969
IUPAC Name propane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(CC(O)=O)C(O)=O

Specifications

Melting Point 159°C
Color White
Quantity 100 g
Formula Weight 176.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3-Propanetricarboxylic Acid
EINECSNumber (2)-4123
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.