Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Trichloropropane, 98+%, Thermo Scientific™

Catalog No. AC149550010
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-18-4
Molecular Formula C3H5Cl3
Molecular Weight (g/mol) 147.42
MDL Number MFCD00000946
InChI Key CFXQEHVMCRXUSD-UHFFFAOYSA-N
Synonym trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132
PubChem CID 7285
ChEBI CHEBI:34036
IUPAC Name 1,2,3-trichloropropane
SMILES ClCC(Cl)CCl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3860g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.4830 to 1.4850 (20°C, 589nm)
Linear Formula ClCH2CH(Cl)CH2Cl
Quantity 1 L
Beilstein 01, 106
Formula Weight 147.43
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 1, 2, 3-Trichloropropane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause cancer.
May damage fertility.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.