Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Trifluorobenzene, 98+%

Catalog No. p-7026193
Click to view available options
Quantity:
1 g
5 g

CAS: 1489-53-8 | C6H3F3 | 132.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1489-53-8
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.085
MDL Number MFCD00012013
InChI Key AJKNNUJQFALRIK-UHFFFAOYSA-N
Synonym trifluorobenzene, benzene, trifluoro, benzene, 1,2,3-trifluoro, 3,4,5-trifluoro-benzene, pubchem1067, 1,2,3-fluorobenzene, 1,2,3-trifluorobenze, 1,2,3-trifluorbenzen, 1,2,6-trifluorobenzene, 2,3,4-trifluorobenzene
PubChem CID 15147
IUPAC Name 1,2,3-trifluorobenzene
SMILES C1=CC(=C(C(=C1)F)F)F

Specifications

Density 1.28
Boiling Point 94°C to 95°C
Flash Point −3°C (26°F)
Refractive Index 1.422
Quantity 1 g
UN Number UN2387
Beilstein 1862388
Formula Weight 132.09
Percent Purity ≥98%
Chemical Name or Material 1,2,3-Trifluorobenzene
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROBENZENE
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.