Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Trimethylbenzene 80.0+%, TCI America™
SDP

Catalog No. T046825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T046825ML 25 mL
T04685ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T046825ML Supplier TCI America Supplier No. T046825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 526-73-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008520
InChI Key FYGHSUNMUKGBRK-UHFFFAOYSA-N
Synonym hemimellitene, hemellitol, hemimellitol, trimethylbenzene, benzene, 1,2,3-trimethyl, benzene, trimethyl, unii-zk4r7uph6r, 1,2,3-trimethylbenzene, zk4r7uph6r, 1,3-trimethylbenzene
PubChem CID 10686
ChEBI CHEBI:34037
IUPAC Name 1,2,3-trimethylbenzene
SMILES CC1=C(C(=CC=C1)C)C

Specifications

Color Yellow
Boiling Point 176°C
Quantity 25 mL
UN Number 3295
Formula Weight 120.20
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3-Trimethylbenzene
EINECSNumber (3)-0007&(3)-3427
RTECSNumber DC3300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.